Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Marimastat 98%

Supplier: AMBEED, INC
Warning

A573351-1MG A573351-5MG A573351-100MG A573351-50MG
76878-756EA 24 USD InStock
76878-756 76878-758 77673-542 77673-544
Marimastat 98%
Marimastat
Precaución: for Research Use Only


Formula: C₁₅H₂₉N₃O₅
MDL Number: MFCD00866242
CAS Number: 154039-60-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty