Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Bromoethoxy)benzeneboronic acid pinacol ester 96%

Supplier: AMBEED, INC
Warning

A573933-100MG A573933-1G A573933-250MG
76878-762EA 19 USD InStock
76878-762 76878-764 76878-766
4-(2-Bromoethoxy)benzeneboronic acid pinacol ester 96%
4-(2-Bromoethoxy)benzeneboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₂₀BBrO₃
MW: 327.03 g/mol
MDL Number: MFCD09027285
CAS Number: 913836-27-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty