Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1-(3-chlorophenyl)-1H-pyrazole 96%

Supplier: AMBEED, INC
Warning

A584567-1G A584567-250MG A584567-5G
76878-840EA 13 USD InStock
76878-840 76878-842 76878-844
4-Bromo-1-(3-chlorophenyl)-1H-pyrazole 96%
4-Bromo-1-(3-chlorophenyl)-1H-pyrazole
Precaución: for Research Use Only


Formula: C₉H₆BrClN₂
MDL Number: MFCD09475849
CAS Number: 957034-94-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty