Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,1,3-Benzothiadiazol-4-ylamine 98%

Supplier: AMBEED, INC
Warning

A598670-1G A598670-250MG A598670-5G
76878-926EA 32 USD InStock
76878-926 76878-928 76878-930
2,1,3-Benzothiadiazol-4-ylamine 98%
2,1,3-Benzothiadiazol-4-ylamine
Precaución: for Research Use Only


Formula: C₆H₅N₃S
MDL Number: MFCD00005810
CAS Number: 767-64-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty