Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ureidobenzeneboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A604631-1G A604631-250MG A604631-5G
76878-966EA 95 USD InStock
76878-966 76878-968 76878-970
4-Ureidobenzeneboronic acid pinacol ester 98%
4-Ureidobenzeneboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₉BN₂O₃
MW: 262.12 g/mol
MDL Number: MFCD06795683
CAS Number: 877134-77-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty