Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-methylphenylboronic acid 96%

Supplier: AMBEED, INC
Warning

A608707-250MG A608707-5G
76879-014EA 103.97 USD InStock
76879-014 76879-016 76879-018
2-Chloro-3-methylphenylboronic acid 96%
2-Chloro-3-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₈BClO₂
MW: 170.4000 g/mol
MDL Number: MFCD06797215
CAS Number: 915070-53-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty