Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Hydroxyethylamino)-5-pyrimidineboronic acid pinacol ester 95%

Supplier: AMBEED, INC
Warning

A609508-100MG A609508-1G A609508-250MG A609508-5G
76879-024EA 37 USD InStock
76879-024 76879-026 76879-028 76879-030
2-(2-Hydroxyethylamino)-5-pyrimidineboronic acid pinacol ester 95%
2-(2-Hydroxyethylamino)-5-pyrimidineboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₂H₂₀BN₃O₃
MDL Number: MFCD12546562
CAS Number: 1218789-34-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty