Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[N-Ethyl-N-(4-methoxybenzyl)sulfamoyl]benzeneboronic acid 98%

Supplier: AMBEED, INC
Warning

A613281-1G A613281-250MG A613281-5G
76879-058EA 163 USD InStock
76879-058 76879-060 76879-062
4-[N-Ethyl-N-(4-methoxybenzyl)sulfamoyl]benzeneboronic acid 98%
4-[N-Ethyl-N-(4-methoxybenzyl)sulfamoyl]benzeneboronic acid
Precaución: for Research Use Only


Formula: C₁₆H₂₀BNO₅S
MDL Number: MFCD08689489
CAS Number: 913835-55-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty