Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Neopentylboronic acid 95%

Supplier: AMBEED, INC
Warning

A614204-10G A614204-1G A614204-250MG A614204-25G
76879-064EA 57 USD InStock
76879-064 76879-066 76879-068 76879-070
Neopentylboronic acid 95%
Neopentylboronic acid
Precaución: for Research Use Only


Formula: C₅H₁₃BO₂
MDL Number: MFCD09750464
CAS Number: 701261-35-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty