Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3-Pyridinyl)-1-propanamine 95%

Supplier: AMBEED, INC
Danger

A621219-10G A621219-1G A621219-250MG
76879-122EA 481 USD InStock
76879-122 76879-124 76879-126
1-(3-Pyridinyl)-1-propanamine 95%
1-(3-Pyridinyl)-1-propanamine
Precaución: for Research Use Only


Formula: C₈H₁₂N₂
MDL Number: MFCD09802326
CAS Number: 60289-67-0
UN: 2810
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty