Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylsulfonylbenzylamine 96%

Supplier: AMBEED, INC
Warning

A621306-1G A621306-250MG
76879-128EA 20 USD InStock
76879-128 77673-682
4-Methylsulfonylbenzylamine 96%
4-Methylsulfonylbenzylamine
Precaución: for Research Use Only


Formula: C₈H₁₁NO₂S
MDL Number: MFCD06213173
CAS Number: 4393-16-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty