Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromobenzaldehyde diethyl acetal 97%

Supplier: AMBEED, INC
Warning

A628434-25G A628434-5G A628434-100G
76879-174EA 28 USD InStock
76879-174 76879-176 77673-704
3-Bromobenzaldehyde diethyl acetal 97%
3-Bromobenzaldehyde diethyl acetal
Precaución: for Research Use Only


Formula: C₁₁H₁₅BrO₂
MW: 259.14 g/mol
MDL Number: MFCD01075697
CAS Number: 75148-49-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty