Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dichloro-5-nitro-2-(propylthio)pyrimidine 97%

Supplier: AMBEED, INC
Warning

A633883-100G A633883-10G A633883-1G A633883-25G A633883-5G
76879-212EA 90 USD InStock
76879-212 76879-214 76879-216 76879-218 76879-220
4,6-Dichloro-5-nitro-2-(propylthio)pyrimidine 97%
4,6-Dichloro-5-nitro-2-(propylthio)pyrimidine
Precaución: for Research Use Only


Formula: C₇H₇Cl₂N₃O₂S
MW: 268.12 g/mol
MDL Number: MFCD10698669
CAS Number: 145783-14-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty