Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-2,3-dihydro-1H-inden-1-ol 95%

Supplier: AMBEED, INC
Warning

A641134-100MG A641134-1G A641134-250MG A641134-5G
76879-254EA 9 USD InStock
76879-254 76879-256 76879-258 76879-260
5-Fluoro-2,3-dihydro-1H-inden-1-ol 95%
5-Fluoro-2,3-dihydro-1H-inden-1-ol
Precaución: for Research Use Only


Formula: C₉H₉FO
MDL Number: MFCD11219501
CAS Number: 52085-92-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty