Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Calcium 4-oxo-4-((2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl)oxy)butanoate 99+%

Supplier: AMBEED, INC
Warning

A645840-100G A645840-25G A645840-5G
76879-292EA 88 USD InStock
76879-292 76879-294 76879-296
Calcium 4-oxo-4-((2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl)oxy)butanoate 99+%
Calcium 4-oxo-4-((2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl)oxy)butanoate
Precaución: for Research Use Only


Formula: C₆₆H₁₀₆CaO₁₀
MW: 1099.61 g/mol
CAS Number: 14638-18-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty