Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-(4-Boronophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 98%

Supplier: AMBEED, INC
Warning

A647030-100G A647030-25G A647030-5G
76879-298EA 430 USD InStock
76879-298 76879-300 76879-302 76879-304
(S)-3-(4-Boronophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 98%
(S)-3-(4-Boronophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid
Precaución: for Research Use Only


Formula: C₁₄H₂₀BNO₆
MDL Number: MFCD01860642
CAS Number: 119771-23-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty