Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyano-4-(trifluoromethyl)benzeneboronic acid neopentyl glycol ester 95%

Supplier: AMBEED, INC
Warning

A653475-1G A653475-250MG A653475-5G
76879-352EA 207 USD InStock
76879-352 76879-354 76879-356
2-Cyano-4-(trifluoromethyl)benzeneboronic acid neopentyl glycol ester 95%
2-Cyano-4-(trifluoromethyl)benzeneboronic acid neopentyl glycol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₃BF₃NO₂
MW: 283.054 g/mol
MDL Number: MFCD08689554
CAS Number: 883898-98-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty