Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1-fluoro-2-naphthaleneboronic acid 98%

Supplier: AMBEED, INC
Warning

A660980-100MG A660980-1G A660980-250MG A660980-5G
76879-394EA 30 USD InStock
76879-394 76879-396 76879-398 76879-400
4-Bromo-1-fluoro-2-naphthaleneboronic acid 98%
4-Bromo-1-fluoro-2-naphthaleneboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₇BBrFO₂
MW: 268.8748 g/mol
MDL Number: MFCD09027236
CAS Number: 913836-09-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty