Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-fluoro-3-nitrobenzoic acid 95%

Supplier: AMBEED, INC
Warning

A666850-1G A666850-250MG A666850-5G
76879-446EA 17 USD InStock
76879-446 76879-448 76879-450
2-Bromo-5-fluoro-3-nitrobenzoic acid 95%
2-Bromo-5-fluoro-3-nitrobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃BrFNO₄
MW: 264.005 g/mol
MDL Number: MFCD12173018
CAS Number: 1055331-73-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty