Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Gly-OPfp 98% (by HPLC)

Supplier: AMBEED, INC
Warning

A671363-1G A671363-25G A671363-5G
76879-466EA 33 USD InStock
76879-466 76879-468 76879-470
Fmoc-Gly-OPfp 98% (by HPLC)
Fmoc-Gly-OPfp
Precaución: for Research Use Only


Formula: C₂₃H₁₄F₅NO₄
MDL Number: MFCD00037641
CAS Number: 86060-85-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty