Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3,4-Difluorophenyl)ethanol 98%

Supplier: AMBEED, INC
Warning

A677422-100MG A677422-1G A677422-250MG
76879-498EA 24 USD InStock
76879-498 76879-500 76879-502
2-(3,4-Difluorophenyl)ethanol 98%
2-(3,4-Difluorophenyl)ethanol
Precaución: for Research Use Only


Formula: C₈H₈F₂O
MDL Number: MFCD00143061
CAS Number: 286440-92-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty