Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzyloxy-3-chlorophenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A706891-1G A706891-250MG A706891-5G
76879-656EA 112 USD InStock
76879-656 76879-658 76879-660
2-Benzyloxy-3-chlorophenylboronic acid 97%
2-Benzyloxy-3-chlorophenylboronic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₂BClO₃
MW: 262.5000 g/mol
MDL Number: MFCD13195644
CAS Number: 1217500-57-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty