Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 3-ethoxy-3-oxoprop-1-en-1-olate 95%

Supplier: AMBEED, INC
Warning

A708678-1G A708678-5G
76879-668EA 94 USD InStock
76879-668 76879-670
Sodium 3-ethoxy-3-oxoprop-1-en-1-olate 95%
Sodium 3-ethoxy-3-oxoprop-1-en-1-olate
Precaución: for Research Use Only


Formula: C₅H₇NaO₃
MDL Number: MFCD00234954
CAS Number: 2649365-68-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty