Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(N'-Hydroxycarbamimidoyl)benzeneboronic acid 95%

Supplier: AMBEED, INC
Warning

A708941-100MG A708941-1G A708941-250MG A708941-5G
76879-672EA 25 USD InStock
76879-672 76879-674 76879-676 76879-678
4-(N'-Hydroxycarbamimidoyl)benzeneboronic acid 95%
4-(N'-Hydroxycarbamimidoyl)benzeneboronic acid
Precaución: for Research Use Only


Formula: C₇H₉BN₂O₃
MDL Number: MFCD08436061
CAS Number: 913835-61-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty