Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Bromophenyl)-6-methylimidazo[1,2-a]pyridine 98%

Supplier: AMBEED, INC
Warning

A709841-1G A709841-250MG A709841-5G
76879-680EA 55 USD InStock
76879-680 76879-682 76879-684
2-(4-Bromophenyl)-6-methylimidazo[1,2-a]pyridine 98%
2-(4-Bromophenyl)-6-methylimidazo[1,2-a]pyridine
Precaución: for Research Use Only


Formula: C₁₄H₁₁BrN₂
MW: 287.16 g/mol
MDL Number: MFCD00444784
CAS Number: 1023-01-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty