Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-aminobutyl(methyl)carbamate 97%

Supplier: AMBEED, INC
Warning

A711003-10G A711003-25G A711003-250MG
76879-696EA 327 USD InStock
76879-696 76879-698 77673-950
tert-Butyl-4-aminobutyl(methyl)carbamate 97%
tert-Butyl-4-aminobutyl(methyl)carbamate
Precaución: For research use only.


Formula: C₁₀H₂₂N₂O₂
MW: 202.2900 g/mol
MDL Number: MFCD03001681
CAS Number: 144222-23-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty