Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine 90%

Supplier: AMBEED, INC
Warning

A715932-100G A715932-10G A715932-1G A715932-25G A715932-500G A715932-5G
76879-756EA 50 USD InStock
76879-756 76879-758 76879-760 76879-762 76879-764 76879-766
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine 90%
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine
Precaución: for Research Use Only


Formula: C₁₃H₂₃NOSi
MDL Number: MFCD00674005
CAS Number: 93102-05-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty