Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,5-Dichloro-9,10-anthraquinone 98%

Supplier: AMBEED, INC
Warning

A726492-100G A726492-1G A726492-25G A726492-5G
76879-854EA 20 USD InStock
76879-854 76879-856 76879-858 76879-860
1,5-Dichloro-9,10-anthraquinone 98%
1,5-Dichloro-9,10-anthraquinone
Precaución: for Research Use Only


Formula: C₁₄H₆Cl₂O₂
MW: 277.11 g/mol
Boiling Pt: 401
MDL Number: MFCD00001190
CAS Number: 82-46-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty