Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Chloro-3-fluorophenyl)methanamine 98%

Supplier: AMBEED, INC
Danger

A733621-25G A733621-5G
76879-894EA 118 USD InStock
76879-894 76879-896
(2-Chloro-3-fluorophenyl)methanamine 98%
(2-Chloro-3-fluorophenyl)methanamine
Precaución: for Research Use Only


Formula: C₇H₇ClFN
MDL Number: MFCD10687077
CAS Number: 72235-54-2
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty