Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dibromo-4-nitropyridine 98%

Supplier: AMBEED, INC
Warning

A752417-100MG A752417-1G A752417-250MG
76879-998EA 12 USD InStock
76879-998 76880-000 76880-002
2,5-Dibromo-4-nitropyridine 98%
2,5-Dibromo-4-nitropyridine
Precaución: for Research Use Only


Formula: C₅H₂Br₂N₂O₂
MDL Number: MFCD16876007
CAS Number: 221241-31-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty