Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-N-methylbenzenesulphonamide 98%

Supplier: AMBEED, INC
Warning

A779746-100MG A779746-1G A779746-250MG
76880-148EA 25 USD InStock
76880-148 76880-150 76880-152
3-Bromo-N-methylbenzenesulphonamide 98%
3-Bromo-N-methylbenzenesulphonamide
Precaución: for Research Use Only


Formula: C₇H₈BrNO₂S
MW: 250.11 g/mol
MDL Number: MFCD07363818
CAS Number: 153435-79-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty