Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid 96%

Supplier: AMBEED, INC
Warning

A787022-1G A787022-250MG A787022-5G
76880-194EA 131 USD InStock
76880-194 76880-196 76880-198
4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid 96%
4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₅H₂₀BNO₅
MDL Number: MFCD11504863
CAS Number: 1150114-74-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty