Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-nitrophenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A795079-1G A795079-250MG A795079-5G
76880-232EA 30 USD InStock
76880-232 76880-234 76880-236
4-Chloro-3-nitrophenylboronic acid pinacol ester 98%
4-Chloro-3-nitrophenylboronic acid pinacol ester


Formula: C₁₂H₁₅BClNO₄
MDL Number: MFCD08235086
CAS Number: 913836-26-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty