Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 6-amino-3-bromopicolinate 98%

Supplier: AMBEED, INC
Warning

A816845-1G A816845-250MG A816845-5G
76880-352EA 47 USD InStock
76880-352 76880-354 76880-356
Ethyl 6-amino-3-bromopicolinate 98%
Ethyl 6-amino-3-bromopicolinate
Precaución: for Research Use Only


Formula: C₈H₉BrN₂O₂
MDL Number: MFCD13182435
CAS Number: 1214332-35-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty