Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-methylbenzeneboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A855161-1G A855161-25G A855161-5G
76880-582EA 21 USD InStock
76880-582 76880-584 76880-586
4-Fluoro-3-methylbenzeneboronic acid pinacol ester 98%
4-Fluoro-3-methylbenzeneboronic acid pinacol ester


Formula: C₁₃H₁₈BFO₂
MDL Number: MFCD18729904
CAS Number: 1001200-60-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty