Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Cys(StBu)-OH 95%

Supplier: AMBEED, INC
Warning

A864079-100MG A864079-10G A864079-1G A864079-250MG A864079-25G A864079-5G
76880-618EA 12 USD InStock
76880-618 76880-620 76880-622 76880-624 76880-626 76880-628
Fmoc-Cys(StBu)-OH 95%
Fmoc-Cys(StBu)-OH
Precaución: for Research Use Only


Formula: C₂₂H₂₅NO₄S₂
MW: 431.565 g/mol
MDL Number: MFCD00042617
CAS Number: 73724-43-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty