Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-N-methylbenzamide 98%

Supplier: AMBEED, INC
Warning

A868422-10G A868422-1G A868422-250MG A868422-25G A868422-5G
76880-654EA 60 USD InStock
76880-654 76880-656 76880-658 76880-660 76880-662
4-Fluoro-N-methylbenzamide 98%
4-Fluoro-N-methylbenzamide
Precaución: for Research Use Only


Formula: C₈H₈FNO
MW: 153.1536 g/mol
MDL Number: MFCD00032531
CAS Number: 701-49-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty