Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxypyrazolo[1,5-a]pyridine 95%

Supplier: AMBEED, INC
Warning

A875641-100MG A875641-1G A875641-250MG
76880-690EA 36 USD InStock
76880-690 76880-692 76880-694
5-Methoxypyrazolo[1,5-a]pyridine 95%
5-Methoxypyrazolo[1,5-a]pyridine
Precaución: for Research Use Only


Formula: C₈H₈N₂O
MDL Number: MFCD16619761
CAS Number: 877994-06-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty