Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Aminomethyl)-4-bromonaphthalene hydrochloride 98%

Supplier: AMBEED, INC
Warning

A893799-1G A893799-250MG A893799-5G
76880-758EA 74 USD InStock
76880-758 76880-760 76880-762
1-(Aminomethyl)-4-bromonaphthalene hydrochloride 98%
1-(Aminomethyl)-4-bromonaphthalene hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₁BrClN
MW: 272.5687 g/mol
MDL Number: MFCD08706223
CAS Number: 578029-09-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty