Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 2-(2-amino-3-(4-bromobenzoyl)phenyl)acetate hydrate(2:2:3) 99%

Supplier: AMBEED, INC
Warning

A895934-1G A895934-250MG A895934-5G
76880-778EA 59 USD InStock
76880-778 76880-780 77674-330
Sodium 2-(2-amino-3-(4-bromobenzoyl)phenyl)acetate hydrate(2:2:3) 99%
Sodium 2-(2-amino-3-(4-bromobenzoyl)phenyl)acetate hydrate(2:2:3)
Precaución: for Research Use Only


Formula: C₁₅H₁₃BrNNaO₄
MDL Number: MFCD09953182
CAS Number: 120638-55-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty