Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-(4-chlorobenzyloxy)-5-methylphenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A907516-100MG A907516-1G A907516-250MG
76880-814EA 11 USD InStock
76880-814 76880-816 76880-818
3-Bromo-2-(4-chlorobenzyloxy)-5-methylphenylboronic acid 97%
3-Bromo-2-(4-chlorobenzyloxy)-5-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₃BBrClO₃
MDL Number: MFCD06411365
CAS Number: 849052-18-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty