Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-6-(4-methyl-1-imidazolyl)pyridazine 98%

Supplier: AMBEED, INC
Warning

A915652-10G A915652-1G A915652-250MG A915652-5G
76880-858EA 190 USD InStock
76880-858 76880-860 76880-862 76880-864
3-Chloro-6-(4-methyl-1-imidazolyl)pyridazine 98%
3-Chloro-6-(4-methyl-1-imidazolyl)pyridazine
Precaución: for Research Use Only


Formula: C₈H₇ClN₄
MDL Number: MFCD09800928
CAS Number: 941294-26-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty