Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-nitrotoluene 95%

Supplier: AMBEED, INC
Warning

A951741-1000G A951741-100G A951741-25G A951741-500G A951741-5G
76880-942EA 167 USD InStock
76880-942 76880-944 76880-946 76880-948 76880-950
2-Chloro-6-nitrotoluene 95%
2-Chloro-6-nitrotoluene
Precaución: for Research Use Only


Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 238
MDL Number: MFCD00007205
CAS Number: 83-42-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty