Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Chloropyrazolo[1,5-a]pyrimidine-3-carbonitrile 95%

Supplier: AMBEED, INC
Warning

A956480-100MG A956480-1G A956480-250MG
76880-968EA 110 USD InStock
76880-968 76880-970 76880-972
7-Chloropyrazolo[1,5-a]pyrimidine-3-carbonitrile 95%
7-Chloropyrazolo[1,5-a]pyrimidine-3-carbonitrile
Precaución: for Research Use Only


Formula: C₇H₃ClN₄
MDL Number: MFCD11109843
CAS Number: 1159982-97-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty