Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl acetylenedicarboxylate 98%

Supplier: AMBEED, INC
Danger

A958456-10G A958456-1G A958456-500G A958456-5G
76880-984EA 9 USD InStock
76880-984 76880-986 76880-988 76880-990
Diethyl acetylenedicarboxylate 98%
Diethyl acetylenedicarboxylate
Precaución: for Research Use Only


Formula: C₈H₁₀O₄
MW: 170.16 g/mol
MDL Number: MFCD00009186
CAS Number: 762-21-0
UN: 1760
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty