Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-(diethylcarbamoyl)phenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A981983-100MG A981983-1G A981983-250MG
76881-048EA 18 USD InStock
76881-048 76881-050 76881-052
3-Chloro-5-(diethylcarbamoyl)phenylboronic acid 98%
3-Chloro-5-(diethylcarbamoyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₅BClNO₃
MDL Number: MFCD09800862
CAS Number: 957120-59-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty