Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-methoxypyridine-4-carboxylic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

20467-1G 20467-500MG 20467-5G
76897-994EA 795.68 USD InStock
76897-994 76897-996 76897-998
2-Fluoro-5-methoxypyridine-4-carboxylic acid ≥95% research grade
2-Fluoro-5-methoxypyridine-4-carboxylic acid


Formula: C₇H₆FNO₃
Storage Temperature: Ambient
MDL Number: MFCD16611205
CAS Number: 1211587-56-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty