Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-(1-hydroxycyclobutyl)acetate ≥95%

Supplier: AOB CHEM USA
Warning

55621-1G 55621-25G 55621-500MG 55621-5G
76943-278EA 195.21 USD InStock
76943-278 76943-280 76943-282 76943-284
Methyl 2-(1-hydroxycyclobutyl)acetate ≥95%
Methyl 2-(1-hydroxycyclobutyl)acetate


Formula: C₇H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD30334949
CAS Number: 1692197-94-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty