Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4-(2,2-Dithoxy-Ethylsulfanyl)-Benzene ≥97%

Supplier: AOB CHEM USA
Warning

21537-1G 21537-25G 21537-500MG 21537-5G
76884-790EA 233.4 USD InStock
76884-790 76884-792 76884-794 76884-796
1-Bromo-4-(2,2-Dithoxy-Ethylsulfanyl)-Benzene ≥97%
1-Bromo-4-(2,2-Dithoxy-Ethylsulfanyl)-Benzene


Formula: C₁₂H₁₇BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD09955275
CAS Number: 96804-05-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty