Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Dibromo-2-(difluoromethoxy)benzene ≥97%

Supplier: AOB CHEM USA
Warning

50602-1G 50602-25G 50602-500MG 50602-5G
76890-944EA 385.11 USD InStock
76890-944 76890-946 76890-948 76890-950
1,4-Dibromo-2-(difluoromethoxy)benzene ≥97%
1,4-Dibromo-2-(difluoromethoxy)benzene


Formula: C₇H₄Br₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD09878215
CAS Number: 1000575-26-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty